Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ube2cbp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_027394.2 | NP_081670.1 | 1 - 2 | 179 | 62 | |
GenBank mRNA | AK011446.1 | - | 1 - 2 | 196 | 62 | |
AK049229.1 | - | 1 - 2 | 180 | 62 | ||
BC115686.1 | - | 1 - 2 | 148 | 40104268 | 62 | |
BC115687.1 | - | 1 - 2 | 147 | 40104269 | 62 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |