Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Apc2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_011789.2 | NP_035919.2 | 7 - 8 | 833 | 88 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Apoptosis Detect Genomic DNA Mus musculus Probe spans exons |