Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cluap1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_029738.2 | NP_084014.1 | 6 - 7 | 700 | 93 | |
GenBank mRNA | AK011846.1 | - | 7 - 8 | 728 | 93 | |
AK134648.1 | - | 6 - 7 | 667 | 93 | ||
AK166050.1 | - | 6 - 7 | 781 | 93 | ||
BC024910.1 | - | 6 - 7 | 700 | 3603454 | 93 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |