Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Phldb3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001102613.1 | NP_001096083.1 | 6 - 7 | 974 | 60 | |
GenBank mRNA | BC052555.1 | - | 1 - 2 | 278 | 60 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |