Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Txndc11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_029582.2 | NP_083858.1 | 2 - 3 | 475 | 97 | |
GenBank mRNA | AK013044.1 | - | 2 - 3 | 523 | 97 | |
AK029166.1 | - | 2 - 3 | 594 | 97 | ||
AK139620.1 | - | 2 - 3 | 557 | 97 | ||
AK155844.1 | - | 2 - 3 | 529 | 97 | ||
AK156154.1 | - | 2 - 3 | 516 | 97 | ||
AK156590.1 | - | 2 - 3 | 555 | 97 | ||
AK157288.1 | - | 2 - 3 | 449 | 97 | ||
AK157948.1 | - | 2 - 3 | 555 | 97 | ||
AK159484.1 | - | 2 - 3 | 532 | 97 | ||
AK170352.1 | - | 2 - 3 | 516 | 97 | ||
BC020363.1 | - | 2 - 3 | 564 | 97 | ||
BC029643.1 | - | 2 - 3 | 475 | 4010810 | 97 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |