Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cdc45 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001161623.1 | NP_001155095.1 | 14 - 15 | 1371 | 93 | |
NM_009862.2 | NP_033992.2 | 16 - 17 | 1748 | 93 | ||
GenBank mRNA | AF081536.1 | - | 16 - 17 | 1748 | 93 | |
AF081537.1 | - | 15 - 16 | 1447 | 93 | ||
AF081538.1 | - | 15 - 16 | 1447 | 93 | ||
AF098068.1 | - | 15 - 16 | 1494 | 93 | ||
AJ223729.1 | - | 15 - 16 | 1468 | 93 | ||
AK147013.1 | - | 15 - 16 | 1465 | 93 | ||
AK167850.1 | - | 14 - 15 | 1359 | 93 | ||
AK210447.1 | - | 3 - 4 | 276 | 93 | ||
BC006856.1 | - | 15 - 16 | 1463 | 93 | ||
BC028635.1 | - | 15 - 16 | 1501 | 1262927 | 93 | |
CO040149.1 | - | 4 - 5 | 93 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |