Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Epha6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_007938.2 | NP_031964.2 | 5 - 6 | 1891 | 78 | |
GenBank mRNA | AK032436.1 | - | 1 - 2 | 136 | 78 | |
BC141090.1 | - | 5 - 6 | 1407 | 9055702 | 78 | |
BC145362.1 | - | 6 - 7 | 1527 | 78 | ||
U58332.1 | - | 5 - 6 | 1607 | 78 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Protein Kinase Mammalian Gene Collection Mus musculus Probe spans exons |