Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Epha3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_010140.3 | NP_034270.1 | 13 - 14 | 2461 | 78 | |
GenBank mRNA | AK045219.1 | - | 13 - 14 | 2462 | 78 | |
AK084921.1 | - | 13 - 14 | 2441 | 78 | ||
AK132335.1 | - | 13 - 14 | 2410 | 78 | ||
BC060164.1 | - | 13 - 14 | 2439 | 78 | ||
BC093483.1 | - | 2462 | 78 | |||
BC138641.1 | - | 13 - 14 | 2384 | 8861661 | 78 | |
BC138643.1 | - | 13 - 14 | 2384 | 8861663 | 78 | |
BC145349.1 | - | 13 - 14 | 2381 | 9053905 | 78 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Protein Kinase Mammalian Gene Collection Mus musculus Probe spans exons |