Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kng1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001102411.1 | NP_001095881.1 | 5 - 6 | 819 | 110 | |
NM_001102412.1 | NP_001095882.1 | 5 - 6 | 819 | 110 | ||
NM_023125.3 | NP_075614.1 | 5 - 6 | 819 | 110 | ||
GenBank mRNA | AK005547.1 | - | 5 - 6 | 784 | 110 | |
AY462058.1 | - | 5 - 6 | 699 | 110 | ||
AY660571.1 | - | 5 - 6 | 726 | 110 | ||
BC018158.1 | - | 5 - 6 | 771 | 4162960 | 110 | |
BC108936.1 | - | 5 - 6 | 775 | 40039851 | 110 | |
D84415.1 | - | 5 - 6 | 699 | 110 | ||
D84435.1 | - | 5 - 6 | 699 | 110 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |