Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tbccd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001081368.1 | NP_001074837.1 | 4 - 5 | 1127 | 69 | |
GenBank mRNA | AK039905.1 | - | 4 - 5 | 952 | 69 | |
AK133804.1 | - | 4 - 5 | 1068 | 69 | ||
BC029679.1 | - | 511 | 69 | |||
BC082558.1 | - | 4 - 5 | 1096 | 30605020 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |