Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Klhl6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_183390.3 | NP_899246.2 | 4 - 5 | 1189 | 82 | |
GenBank mRNA | AY350736.1 | - | 4 - 5 | 1117 | 82 | |
BC100568.1 | - | 4 - 5 | 1176 | 30073940 | 82 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |