Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Usp28 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_175482.3 | NP_780691.2 | 4 - 5 | 474 | 71 | |
GenBank mRNA | AK166511.1 | - | 4 - 5 | 469 | 71 | |
BB659010.1 | - | 4 - 5 | 441 | 71 | ||
BC066033.1 | - | 4 - 5 | 445 | 6835629 | 71 | |
BC088733.1 | - | 4 - 5 | 396 | 30459522 | 71 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |