Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0900001693.mm.1 |
NM_178415.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900001693.mm.1 |
NM_178415.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900001693.mm.1 |
NM_181316.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900001693.mm.1 |
NM_181316.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900001693.mm.1 |
TC0900000223.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0900001701.mm.1 |
NM_178415.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900001701.mm.1 |
NM_178415.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900001701.mm.1 |
NM_181316.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900001701.mm.1 |
NM_181316.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900001701.mm.1 |
TC0900000223.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0900001700.mm.1 |
NM_178415.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900001700.mm.1 |
NM_178415.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900001700.mm.1 |
NM_181316.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900001700.mm.1 |
NM_181316.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900001700.mm.1 |
TC0900000223.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0900001695.mm.1 |
NM_178415.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900001695.mm.1 |
NM_178415.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900001695.mm.1 |
NM_181316.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900001695.mm.1 |
NM_181316.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900001695.mm.1 |
TC0900000223.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0900001697.mm.1 |
NM_178415.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900001697.mm.1 |
NM_178415.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900001697.mm.1 |
TC0900000223.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0900001694.mm.1 |
NM_178415.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900001694.mm.1 |
NM_178415.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900001694.mm.1 |
NM_181316.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900001694.mm.1 |
NM_181316.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900001694.mm.1 |
TC0900000223.mm.1 |
MTA TC |