Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bbs9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_178415.1 | NP_848502.1 | 8 - 9 | 1114 | 79 | |
NM_181316.4 | NP_851833.2 | 7 - 8 | 1058 | 79 | ||
GenBank mRNA | AK028200.1 | - | 7 - 8 | 1148 | 79 | |
AK086114.1 | - | 7 - 8 | 1030 | 79 | ||
AK144730.1 | - | 8 - 9 | 1085 | 79 | ||
AK166297.1 | - | 7 - 8 | 1037 | 79 | ||
BC030419.1 | - | 4 - 5 | 407 | 79 | ||
BC046426.1 | - | 7 - 8 | 1070 | 3601505 | 79 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |