Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Chrd | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001278041.1 | NP_001264970.1 | 19 - 20 | 2527 | 77 | |
NM_009893.2 | NP_034023.1 | 19 - 20 | 2461 | 77 | ||
GenBank mRNA | AF069501.1 | - | 19 - 20 | 2461 | 77 | |
AF096276.1 | - | 19 - 20 | 2440 | 77 | ||
AK149445.1 | - | 19 - 20 | 2436 | 77 | ||
BC145017.1 | - | 17 - 18 | 2575 | 77 | ||
BC145018.1 | - | 19 - 20 | 2495 | 77 | ||
BC145019.1 | - | 19 - 20 | 2532 | 77 | ||
BC145696.1 | - | 19 - 20 | 2525 | 40130999 | 77 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |