Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mfi2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_013900.2 | NP_038928.1 | 12 - 13 | 1802 | 64 | |
GenBank mRNA | AB024336.1 | - | 12 - 13 | 1802 | 64 | |
AK036444.1 | - | 12 - 13 | 1737 | 64 | ||
BC040347.1 | - | 12 - 13 | 1725 | 3672916 | 64 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |