Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cep19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_025892.2 | NP_080168.1 | 2 - 3 | 385 | 68 | |
GenBank mRNA | AK005324.1 | - | 2 - 3 | 385 | 68 | |
AK007713.1 | - | 2 - 3 | 369 | 68 | ||
BC023715.1 | - | 1 - 2 | 65 | 5346460 | 68 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |