Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Smad3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_016769.4 | NP_058049.3 | 6 - 7 | 1195 | 67 | |
GenBank mRNA | AB008192.1 | - | 6 - 7 | 904 | 67 | |
AF016189.1 | - | 6 - 7 | 1171 | 67 | ||
AK041743.1 | - | 6 - 7 | 716 | 67 | ||
AK048626.1 | - | 6 - 7 | 1195 | 67 | ||
AK083158.1 | - | 6 - 7 | 803 | 67 | ||
BC066850.1 | - | 6 - 7 | 881 | 30432720 | 67 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |