Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sik2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_178710.3 | NP_848825.2 | 6 - 7 | 926 | 103 | |
GenBank mRNA | AB067780.1 | - | 6 - 7 | 827 | 103 | |
AK090386.1 | - | 6 - 7 | 827 | 103 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Protein Kinase Mus musculus Probe spans exons |