Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lztr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_025808.3 | NP_080084.2 | 13 - 14 | 1499 | 62 | |
GenBank mRNA | AK004561.2 | - | 13 - 14 | 1499 | 62 | |
AK005037.1 | - | 13 - 14 | 1498 | 62 | ||
AK083411.1 | - | 13 - 14 | 1503 | 62 | ||
AK088938.1 | - | 4 - 5 | 431 | 62 | ||
AK132817.1 | - | 13 - 14 | 1498 | 62 | ||
BC034400.1 | - | 13 - 14 | 1493 | 2651456 | 62 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |