Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dennd4a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001162917.1 | NP_001156389.1 | 26 - 27 | 4984 | 90 | |
GenBank mRNA | AK035923.1 | - | 4 - 5 | 1082 | 90 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Mus musculus Probe spans exons |