Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mpzl3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001093749.2 | NP_001087218.1 | 4 - 5 | 683 | 78 | |
GenBank mRNA | EF102773.1 | - | 4 - 5 | 647 | 78 |
Set Membership: |
Amplicon length between 71 and 85 Mus musculus Probe spans exons |