Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zwilch | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_026507.4 | NP_080783.3 | 13 - 14 | 1372 | 121 | |
GenBank mRNA | AK009559.1 | - | 13 - 14 | 1343 | 121 | |
AK019825.1 | - | 13 - 14 | 1350 | 121 | ||
AK050917.1 | - | 13 - 14 | 1338 | 121 | ||
BC027435.1 | - | 13 - 14 | 1320 | 2812472 | 121 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |