Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tipin | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_025372.3 | NP_079648.1 | 6 - 7 | 516 | 130 | |
GenBank mRNA | AK003451.1 | - | 6 - 7 | 510 | 130 | |
AK003802.1 | - | 6 - 7 | 517 | 130 | ||
AK011357.1 | - | 6 - 7 | 511 | 130 | ||
AK038111.1 | - | 515 | 130 | |||
BC016211.1 | - | 5 - 6 | 493 | 4924605 | 130 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |