Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Carm1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_021531.6 | NP_067506.2 | 5 - 6 | 788 | 67 | |
NM_153141.1 | NP_694781.1 | 5 - 6 | 788 | 67 | ||
GenBank mRNA | AF117887.2 | - | 5 - 6 | 698 | 67 | |
AK158242.1 | - | 5 - 6 | 753 | 67 | ||
AK158757.1 | - | 5 - 6 | 628 | 67 | ||
BC036974.1 | - | 5 - 6 | 772 | 4935077 | 67 | |
BC071267.1 | - | 6 - 7 | 904 | 67 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |