Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Adam10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_007399.3 | NP_031425.2 | 5 - 6 | 788 | 109 | |
GenBank mRNA | AF011379.1 | - | 5 - 6 | 609 | 109 | |
BC057346.1 | - | 5 - 6 | 750 | 109 | ||
BC066207.1 | - | 5 - 6 | 793 | 109 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Protease Mammalian Gene Collection Mus musculus Probe spans exons |