Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Adam10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_007399.3 | NP_031425.2 | 7 - 8 | 1032 | 84 | |
GenBank mRNA | AF011379.1 | - | 7 - 8 | 853 | 84 | |
AK036599.1 | - | 1 - 2 | 76 | 84 | ||
BC057346.1 | - | 7 - 8 | 994 | 84 | ||
BC066207.1 | - | 7 - 8 | 1037 | 84 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Protease Mammalian Gene Collection Mus musculus 1700 Probe spans exons |