Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Amotl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001081395.1 | NP_001074864.1 | 10 - 11 | 2507 | 93 | |
GenBank mRNA | AK016526.1 | - | 11 - 12 | 2367 | 93 | |
AK220238.1 | - | 8 - 9 | 1741 | 93 | ||
AX763684.1 | - | 8 - 9 | 2031 | 93 | ||
BC055117.1 | - | 2 - 3 | 232 | 93 | ||
BC151000.1 | - | 10 - 11 | 2315 | 9087913 | 93 | |
BC151004.1 | - | 10 - 11 | 2315 | 9087917 | 93 | |
BC151007.1 | - | 10 - 11 | 2315 | 9087920 | 93 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |