Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0900014837.mm.1 |
NM_001081395.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900014837.mm.1 |
NM_001081395.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900014837.mm.1 |
TC0900001771.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0900014833.mm.1 |
NM_001081395.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900014833.mm.1 |
NM_001081395.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900014833.mm.1 |
TC0900001771.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0900014832.mm.1 |
NM_001081395.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900014832.mm.1 |
NM_001081395.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900014832.mm.1 |
TC0900001771.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0900014839.mm.1 |
NM_001081395.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900014839.mm.1 |
NM_001081395.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900014839.mm.1 |
TC0900001771.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0900014835.mm.1 |
NM_001081395.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900014835.mm.1 |
NM_001081395.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900014835.mm.1 |
TC0900001771.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0900014834.mm.1 |
NM_001081395.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900014834.mm.1 |
NM_001081395.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900014834.mm.1 |
TC0900001771.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0900014841.mm.1 |
NM_001081395.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900014841.mm.1 |
NM_001081395.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900014841.mm.1 |
TC0900001771.mm.1 |
MTA TC |