Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bex6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001033539.2 | NP_001028711.1 | 1 - 2 | 100 | 183 | |
GenBank mRNA | AK157452.1 | - | 1 - 2 | 101 | 183 | |
AY833559.1 | - | 1 - 2 | 100 | 183 |
Set Membership: |
Amplicon length greater than or equal to 101 Mus musculus Probe spans exons |