Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rcn2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_011992.2 | NP_036122.2 | 6 - 7 | 1023 | 146 | |
GenBank mRNA | AF049125.1 | - | 6 - 7 | 1012 | 146 | |
AK041996.1 | - | 1020 | 146 | |||
AK077486.1 | - | 6 - 7 | 999 | 146 | ||
AK169765.1 | - | 6 - 7 | 997 | 146 | ||
BC132320.1 | - | 6 - 7 | 988 | 40130597 | 146 | |
BC145668.1 | - | 6 - 7 | 988 | 40130971 | 146 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |