Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Atcay | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_178662.3 | NP_848777.1 | 6 - 7 | 988 | 74 | |
GenBank mRNA | AK028298.1 | - | 6 - 7 | 995 | 74 | |
AK038483.1 | - | 6 - 7 | 998 | 74 | ||
AK038990.1 | - | 6 - 7 | 989 | 74 | ||
AK039411.1 | - | 6 - 7 | 996 | 74 | ||
AK162959.1 | - | 6 - 7 | 996 | 74 | ||
AY349150.1 | - | 6 - 7 | 953 | 74 | ||
BC048903.1 | - | 6 - 7 | 972 | 6491141 | 74 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Apoptosis Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |