Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Srl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_175347.4 | NP_780556.2 | 3 - 4 | 1472 | 88 | |
GenBank mRNA | AB113383.1 | - | 3 - 4 | 1472 | 88 | |
AK036394.1 | - | 2 - 3 | 172 | 88 | ||
BC145687.1 | - | 3 - 4 | 1494 | 40130990 | 88 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |