Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Egln2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053208.4 | NP_444438.2 | 1 - 2 | 119 | 53 | |
GenBank mRNA | AF453879.1 | - | 1 - 2 | 41 | 53 | |
AJ310547.1 | - | 1 - 2 | 90 | 53 | ||
AK075582.1 | - | 1 - 2 | 71 | 53 | ||
AK152056.1 | - | 1 - 2 | 80 | 53 | ||
AK152223.1 | - | 1 - 2 | 77 | 53 | ||
AK159206.1 | - | 1 - 2 | 72 | 53 | ||
AK167477.1 | - | 1 - 2 | 73 | 53 | ||
BC023299.1 | - | 1 - 2 | 53 | 5135579 | 53 | |
BC086764.1 | - | 1 - 2 | 58 | 5685311 | 53 | |
CB248412.1 | - | 1 - 2 | 119 | 53 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus 1700 Probe spans exons |