Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Plekhg2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001083912.1 | NP_001077381.1 | 15 - 16 | 2300 | 64 | |
NM_001290542.1 | NP_001277471.1 | 15 - 16 | 2523 | 64 | ||
NM_138752.2 | NP_620091.2 | 15 - 16 | 2303 | 64 | ||
GenBank mRNA | AF465238.1 | - | 15 - 16 | 2768 | 64 | |
AK131110.1 | - | 15 - 16 | 2132 | 64 | ||
AK144152.1 | - | 15 - 16 | 2463 | 64 | ||
BC040403.1 | - | 13 - 14 | 1299 | 64 | ||
BC052436.1 | - | 13 - 14 | 1712 | 6837328 | 64 | |
BC056971.1 | - | 15 - 16 | 1571 | 6821362 | 64 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |