Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Plekhg2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001083912.1 | NP_001077381.1 | 16 - 17 | 2321 | 102 | |
NM_001290542.1 | NP_001277471.1 | 16 - 17 | 2544 | 102 | ||
NM_138752.2 | NP_620091.2 | 16 - 17 | 2324 | 102 | ||
GenBank mRNA | AF465238.1 | - | 16 - 17 | 2789 | 102 | |
AK131110.1 | - | 16 - 17 | 2153 | 102 | ||
AK144152.1 | - | 16 - 17 | 2484 | 102 | ||
BC040403.1 | - | 14 - 15 | 1320 | 102 | ||
BC052436.1 | - | 14 - 15 | 1733 | 6837328 | 102 | |
BC056971.1 | - | 16 - 17 | 1592 | 6821362 | 102 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |