Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gpx4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001037741.3 | NP_001032830.2 | 1 - 2 | 285 | 56 | |
GenBank mRNA | AB072797.1 | - | 1 - 2 | 267 | 56 | |
AF274027.1 | - | 1 - 2 | 261 | 56 | ||
BU961981.1 | - | 288 | 56 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Detect Genomic DNA Mus musculus 1700 Probe spans exons |