Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Prmt2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AF169620.1 | - | 2 - 3 | 140 | 92 | |
AK077456.1 | - | 2 - 3 | 197 | 92 | ||
AK140200.1 | - | 2 - 3 | 224 | 92 | ||
AK144412.1 | - | 2 - 3 | 195 | 92 | ||
AK145829.1 | - | 2 - 3 | 197 | 92 | ||
AK146754.1 | - | 2 - 3 | 196 | 92 | ||
AK146966.1 | - | 2 - 3 | 204 | 92 | ||
AK159186.1 | - | 2 - 3 | 203 | 92 | ||
BC088997.1 | - | 2 - 3 | 154 | 92 | ||
BC122563.1 | - | 1 - 2 | 128 | 40107557 | 92 | |
BC125275.1 | - | 1 - 2 | 128 | 40107558 | 92 | |
BY288156.1 | - | 2 - 3 | 232 | 92 | ||
BY727303.1 | - | 2 - 3 | 223 | 92 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |