Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Vps26a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001113355.1 | NP_001106826.1 | 1 - 2 | 219 | 133 | |
GenBank mRNA | AK028096.1 | - | 1 - 2 | 220 | 133 |
Set Membership: |
Amplicon length greater than or equal to 101 Mus musculus Probe spans exons |