Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pcbp3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | BC042440.1 | - | 1 - 2 | 67 | 4511401 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |