Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Smarcb1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001161853.1 | NP_001155325.1 | 2 - 3 | 426 | 107 | |
GenBank mRNA | AB017344.1 | - | 2 - 3 | 207 | 107 | |
AK145552.1 | - | 2 - 3 | 415 | 107 | ||
AK150169.1 | - | 2 - 3 | 427 | 107 |
Set Membership: |
Amplicon length greater than or equal to 101 Mus musculus Probe spans exons |