Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Jsrp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_028001.3 | NP_082277.2 | 3 - 4 | 199 | 76 | |
GenBank mRNA | AK009016.1 | - | 3 - 4 | 194 | 76 | |
BC104136.1 | - | 2 - 3 | 113 | 76 | ||
BC104137.1 | - | 2 - 3 | 114 | 76 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |