Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mcm3ap | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_019434.2 | NP_062307.2 | 27 - 28 | 5957 | 64 | |
GenBank mRNA | AK122313.1 | - | 27 - 28 | 6001 | 64 | |
AK165254.1 | - | 16 - 17 | 2662 | 64 | ||
BC052452.1 | - | 27 - 28 | 5957 | 6409827 | 64 | |
BC067414.1 | - | 26 - 27 | 4892 | 64 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |