Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nde1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001114085.1 | NP_001107557.1 | 4 - 5 | 525 | 67 | |
NM_001285503.1 | NP_001272432.1 | 4 - 5 | 525 | 67 | ||
NM_023317.2 | NP_075806.2 | 4 - 5 | 525 | 67 | ||
GenBank mRNA | AF322073.1 | - | 3 - 4 | 432 | 67 | |
AK012830.1 | - | 4 - 5 | 455 | 67 | ||
AK012861.1 | - | 4 - 5 | 475 | 67 | ||
AK037289.1 | - | 4 - 5 | 468 | 67 | ||
AK146982.1 | - | 4 - 5 | 455 | 67 | ||
AK150826.1 | - | 4 - 5 | 515 | 67 | ||
AK169743.1 | - | 4 - 5 | 455 | 67 | ||
BC023267.1 | - | 4 - 5 | 504 | 3672565 | 67 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |