Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pml | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_008884.5 | NP_032910.3 | 2 - 3 | 709 | 102 | |
NM_178087.4 | NP_835188.2 | 2 - 3 | 709 | 102 | ||
GenBank mRNA | AK028044.1 | - | 2 - 3 | 711 | 102 | |
AK052305.1 | - | 2 - 3 | 709 | 102 | ||
AK054135.1 | - | 2 - 3 | 709 | 102 | ||
AK156279.1 | - | 2 - 3 | 591 | 102 | ||
BC020990.1 | - | 2 - 3 | 654 | 4188386 | 102 | |
KJ650238.1 | - | 2 - 3 | 613 | 102 | ||
U33626.2 | - | 2 - 3 | 606 | 102 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |