Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fxyd2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_052823.2 | NP_439888.1 | 2 - 3 | 200 | 109 | |
GenBank mRNA | AA396010.1 | - | 2 - 3 | 148 | 108 | |
AY626244.1 | - | 2 - 3 | 200 | 109 | ||
BC069892.1 | - | 2 - 3 | 124 | 109 | ||
CB600439.1 | - | 2 - 3 | 160 | 109 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Transporter Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |