Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ubl7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001122873.1 | NP_001116345.1 | 8 - 9 | 818 | 69 | |
NM_027086.3 | NP_081362.2 | 8 - 9 | 766 | 69 | ||
GenBank mRNA | AK154496.1 | - | 8 - 9 | 776 | 69 | |
AK179100.1 | - | 4 - 5 | 330 | 69 | ||
AK183337.1 | - | 4 - 5 | 330 | 69 | ||
AK207226.1 | - | 4 - 5 | 330 | 69 | ||
AK209878.1 | - | 4 - 5 | 350 | 69 | ||
AK212304.1 | - | 4 - 5 | 331 | 69 | ||
AK214469.1 | - | 2 - 3 | 178 | 69 | ||
BC016456.1 | - | 7 - 8 | 752 | 4912715 | 69 | |
BC094047.1 | - | 7 - 8 | 755 | 5253101 | 69 | |
EL608156.1 | - | 3 - 4 | 69 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |