Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cntn5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001033359.2 | NP_001028531.2 | 4 - 5 | 709 | 86 | |
NM_001170787.1 | NP_001164258.1 | 8 - 9 | 1373 | 86 | ||
GenBank mRNA | AK138164.1 | - | 4 - 5 | 710 | 86 | |
AK162856.1 | - | 4 - 5 | 709 | 86 |
Set Membership: |
Amplicon length between 86 and 100 Mus musculus Probe spans exons |