Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Trip4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001170907.1 | NP_001164378.1 | 8 - 9 | 94 | ||
NM_019797.4 | NP_062771.2 | 11 - 12 | 94 | |||
GenBank mRNA | AF197574.1 | - | 8 - 9 | 1191 | 94 | |
AF539614.1 | - | 11 - 12 | 2015 | 94 | ||
AK039024.1 | - | 8 - 9 | 94 | |||
AK146298.1 | - | 8 - 9 | 1199 | 94 | ||
BC021316.1 | - | 8 - 9 | 1204 | 5027899 | 94 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |