Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc24a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_144813.1 | NP_659062.1 | 5 - 6 | 2296 | 122 | |
GenBank mRNA | BC016094.1 | - | 5 - 6 | 2296 | 4504496 | 122 |
BC044859.1 | - | 4 - 5 | 1732 | 122 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Transporter Mammalian Gene Collection Mus musculus Probe spans exons |